1-Ethyl-3-methylimidazoliumtetrafluoroborate, (max. 3% H₂O) 98+% (dry weight)
Supplier: Thermo Scientific
Warning
Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium tetrafluoroborate
L19763.04 L19763.09 L19763.18
L19763.04EA 53.9 EUR
L19763.04 L19763.09 L19763.18
1-Ethyl-3-methylimidazoliumtetrafluoroborate, (max. 3% H₂O) 98+% (dry weight)
1-Ethyl-3-methylimidazolium tetrafluoroborate
Purity: 98+% CAS No.: 143314-16-3 Molecular Formula: C6H11BF4N2 Formula Weight: 197.98 Flash Point: None Refractive Index: 1.4100 Melting Point: 15° MDL No.: MFCD00216668
Formula: C₆H₁₁BF₄N₂
MW: 197.97 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: 15 °C
Density: 1.280
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00216668
CAS Number: 143314-16-3