1-(3-Methoxyphenyl)-2-thiourea
Supplier: Thermo Scientific
Warning
Synonyms: 3-Methoxyphenylthiourea, 1-(3-Methoxyphenyl)thiourea, N-(3-Methoxyphenyl)thiourea
AW01294.250 AW01294.1
MAYBAW01294.250EA 0 EUR
MAYBAW01294.250 MAYBAW01294.1
1-(3-Methoxyphenyl)-2-thiourea
1-(3-Methoxyphenyl)-2-thiourea
Formula: (C₆H₄(OCH₃))NC(S)NH₂
MW: 182.25 g/mol
MDL Number: MFCD00060446
CAS Number: 37014-08-7
UN: 2811
ADR: 6.1,III