Ethyl bromodifluoroacetate
Supplier: Thermo Scientific
Synonyms:
Ethyl 2-bromo-2,2-difluoroacetate, Bromodifluoroacetic ethyl ester
BTB12440.1
BTB12440.10
BTB12440.50
MAYBBTB12440.1EA
0
EUR
MAYBBTB12440.1
MAYBBTB12440.10
MAYBBTB12440.50
Ethyl bromodifluoroacetate
Ethyl bromodifluoroacetate
Formula:
BrF₂CCOOCH₂CH₃ MW: 202.98 g/mol Boiling Pt: 112 Density: 1.5926 g/cm³ (20 °C) |
MDL Number:
MFCD00042069 CAS Number: 667-27-6 EINECS: 211-567-0 UN: 2924 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...