1-Adamantanemethanol

Supplier: Thermo Scientific

Synonyms: 1-(Hydroxymethyl)adamantane, 1-(Hydroxymethyl)tricyclo[3.3.1.1(3,7)]decane, Adamantan-1-ylmethanol, 1-Adamantylmethanol

BTB13915.1 BTB13915.10 BTB13915.50
MAYBBTB13915.1EA 0 EUR
MAYBBTB13915.1 MAYBBTB13915.10 MAYBBTB13915.50
1-Adamantanemethanol
1-Adamantanemethanol
Formula: C₁₁H₁₈O
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 115…118 °C
MDL Number: MFCD00074751
CAS Number: 770-71-8
EINECS: 212-225-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR