3-Bromofuran

Supplier: Thermo Scientific

CC02110.1 CC02110EA CC02110.50
MAYBCC02110.1EA 0 EUR
MAYBCC02110.1 MAYBCC02110EA MAYBCC02110.50
3-Bromofuran
3-Bromofuran
Formula: C₄H₃BrO
MW: 146.97 g/mol
Boiling Pt: 103
Density: 1.6957 g/cm³ (0 °C)
MDL Number: MFCD00005347
CAS Number: 22037-28-1
EINECS: 244-744-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR