3-Pyridylmethanol
Supplier: Thermo Scientific
Warning
Synonyms: 3-(Hydroxymethyl)pyridine, 3-Pyridinemethanol, 3-Pyridylcarbinol, Pyridine-3-methanol, 3-Pyridylmethanol
CC04109.1 CC04109.10 CC04109.50
MAYBCC04109.1EA 0 EUR
MAYBCC04109.1 MAYBCC04109.10 MAYBCC04109.50
3-Pyridylmethanol
3-Pyridylmethanol
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 266
Melting Pt: –7 °C
Density: 1.1357 g/cm³ (20 °C)
MDL Number: MFCD00006407
CAS Number: 100-55-0
EINECS: 202-864-6
Merck Index: 12,06614