4'-(Trifluoromethyl)acetophenone
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-[4-(Trifluoromethyl)phenyl]ethan-1-one, 4-(Trifluoromethyl)acetophenone, p-(Trifluoromethyl)acetophenone
RF03240.1
RF03240.10
RF03240.50
MAYBRF03240.1EA
0
EUR
MAYBRF03240.1
MAYBRF03240.10
MAYBRF03240.50
4'-(Trifluoromethyl)acetophenone
4'-(Trifluoromethyl)acetophenone
Formula:
C₉H₇F₃O MW: 188.15 g/mol Boiling Pt: 205 °C (1013 hPa) Melting Pt: 26…29 °C Density: 0,924 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000401 CAS Number: 709-63-7 EINECS: 211-913-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...