3-Amino-5-methylisoxazole

Supplier: Thermo Scientific

Synonyms: 5-Methylisoxazol-3-amine, 3-Amino-5-methylisoxazol, 5-Methylisoxazol-3-ylamine

SB00621.1 SB00621.10
MAYBSB00621.1EA 0 EUR
MAYBSB00621.1 MAYBSB00621.10
3-Amino-5-methylisoxazole
3-Amino-5-methylisoxazole
Formula: C₄H₆N₂O
MW: 98.1 g/mol
Boiling Pt: 194 °C (1013 hPa)
Melting Pt: 56…60 °C
MDL Number: MFCD00003155
CAS Number: 1072-67-9
EINECS: 214-013-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR