2,5-Diphenyloxazole
Supplier: Thermo Scientific
|
Warning
|
SB00840.1
SB00840.10
SB00840.50
MAYBSB00840.1EA
0
EUR
MAYBSB00840.1
MAYBSB00840.10
MAYBSB00840.50
2,5-Diphenyloxazole
2,5-Diphenyloxazole
Formula:
C₁₅H₁₁NO Boiling Pt: 360 °C (1013 hPa) Melting Pt: 70…72 °C Density: 1,094 g/cm³ (99 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005306 CAS Number: 92-71-7 EINECS: 202-181-3 |