6-Nitroveratraldehyde

Supplier: Thermo Scientific

Synonyms: 4,5-Dimethoxy-2-nitrobenzaldehyde, 3,4-Dimethoxy-6-nitrobenzaldehyde

SB00938.1 SB00938.10 SB00938.50
MAYBSB00938.1EA 0 EUR
MAYBSB00938.1 MAYBSB00938.10 MAYBSB00938.50
6-Nitroveratraldehyde
6-Nitroveratraldehyde
Formula: C₉H₉NO₅
Melting Pt: 134…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007134
CAS Number: 20357-25-9
EINECS: 243-762-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR