2-Acetylthiophene
Supplier: Thermo Scientific
Danger
Synonyms: 1-Thiophen-2-yl-ethanone, 1-(2-Thienyl)-1-ethanone, Methyl 2-thienyl ketone
SB01124.1 SB01124.10 SB01124.50
MAYBSB01124.1EA 0 EUR
MAYBSB01124.1 MAYBSB01124.10 MAYBSB01124.50
2-Acetylthiophene
2-Acetylthiophene
Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.1704 g/cm³ (20 °C)
MDL Number: MFCD00005442
CAS Number: 88-15-3
EINECS: 201-804-6
UN: 2810
ADR: 6.1,II