4-Benzyloxyphenol

Supplier: Thermo Scientific

Synonyms: Monobenzone4-Hydroxyphenyl benzyl etherHydroquinone monobenzyl ether4-(Benzyloxy)phenol

SB01455.1 SB01455.10 SB01455.50
MAYBSB01455.1EA 0 EUR
MAYBSB01455.1 MAYBSB01455.10 MAYBSB01455.50
4-Benzyloxyphenol
4-Benzyloxyphenol
Formula: C₁₃H₁₂O₂
MW: 200.24 g/mol
Boiling Pt: 328
Melting Pt: 117.9…121.5 °C
Density: 1.26 g/cm³ (20 °C)
MDL Number: MFCD00002333
CAS Number: 103-16-2
EINECS: 203-083-3
Merck Index: 12,06331

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR