Ethyl propiolate
Supplier: Thermo Scientific
Synonyms: Ethyl propynoate, 2-Propynoic acid ethyl ester, Ethyl acetylenecarboxylate, Propiolic acid ethyl ester
SB01463.1 SB01463.50
MAYBSB01463.1EA 0 EUR
MAYBSB01463.1 MAYBSB01463.50
Ethyl propiolate
Ethyl propiolate
Formula: C₅H₆O₂
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: 9 °C
Density: 0.9788 g/cm³ (4 °C)
MDL Number: MFCD00009184
CAS Number: 623-47-2
EINECS: 210-795-8
UN: 3272
ADR: 3,III