5-Chloro-2-methoxyphenyl isothiocyanate

Supplier: Thermo Scientific

SB01657DA SB01657EA SB01657.50
MAYBSB01657DAEA 0 EUR
MAYBSB01657DA MAYBSB01657EA MAYBSB01657.50
5-Chloro-2-methoxyphenyl isothiocyanate
Formula: C₈H₆ClNOS
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 61 °C
MDL Number: MFCD00041073
CAS Number: 63429-99-2
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR