3,5-Dichloro-α,α,α-trifluorotoluene
Supplier: Thermo Scientific
SEW04383.250
SEW04383.1
MAYBSEW04383.250EA
0
EUR
MAYBSEW04383.250
MAYBSEW04383.1
3,5-Dichloro-α,α,α-trifluorotoluene
3,5-Dichloro-α,α,α-trifluorotoluene
Formula:
C₇H₃Cl₂F₃ MW: 215 g/mol |
MDL Number:
MFCD00203929 CAS Number: 54773-20-5 EINECS: 259-337-9 UN: 2234 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...