1-(2-Fluorophenyl)-2-thiourea
Supplier: Thermo Scientific
Danger
Synonyms: (2-Fluorophenyl)thiourea, N-(2-Fluorophenyl)thiourea, 2-Fluorophenylthiourea
SPB06700.1 SPB06700.10 SPB06700.50
MAYBSPB06700.1EA 0 EUR
MAYBSPB06700.1 MAYBSPB06700.10 MAYBSPB06700.50
1-(2-Fluorophenyl)-2-thiourea
1-(2-Fluorophenyl)-2-thiourea
Formula: (C₆H₄F)NC(S)NH₂
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 146 °C
Density: 1.423 g/cm³ (20 °C)
MDL Number: MFCD00041148
CAS Number: 656-32-6
UN: 2811
ADR: 6.1,I