3-Iodo-o-xylene
Supplier: Thermo Scientific
Warning
Synonyms: 1,2-Dimethyl-3-iodobenzene, 2,3-Dimethyl-1-iodobenzene, 1-Iodo-2,3-dimethylbenzene, 2,3-Dimethyliodobenzene
TL00125.1 TL00125.10 TL00125.50
MAYBTL00125.1EA 0 EUR
MAYBTL00125.1 MAYBTL00125.10 MAYBTL00125.50
3-Iodo-o-xylene
3-Iodo-o-xylene
Formula: C₈H₉I
MW: 232.06 g/mol
Boiling Pt: 110…111 °C (12 mmHg)
Density: 1,64 g/cm³ (20 °C)
MDL Number: MFCD00041437
CAS Number: 31599-60-7