2,5-Dimethoxybenzylamine

Supplier: Thermo Scientific

Synonyms: 1-(2,5-Dimethoxyphenyl)methanamine

TL00178DA TL00178EA
MAYBTL00178DAEA 0 EUR
MAYBTL00178DA MAYBTL00178EA
2,5-Dimethoxybenzylamine
2,5-Dimethoxybenzylamine
Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
MDL Number: MFCD00052811
CAS Number: 3275-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR