4-Methoxybenzenesulphonyl chloride
Supplier: Thermo Scientific
MAYBTL00513.10EA
0
EUR
MAYBTL00513.10
MAYBTL00513.50
MAYBTL00513.1
4-Methoxybenzenesulphonyl chloride
4-Methoxybenzenesulphonyl chloride
Formula:
C₇H₇ClO₃S MW: 206,65 g/mol Boiling Pt: 316 °C (1013 hPa) Melting Pt: 40…42 °C |
MDL Number:
MFCD00007446 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...