N-Ethyl-4-(trifluoromethoxy)aniline

Supplier: Thermo Scientific

Synonyms: 4-(Trifluoromethoxy)ethylaminobenzene

TL00676DA TL00676EA
MAYBTL00676DAEA 0 EUR
MAYBTL00676DA MAYBTL00676EA
N-Ethyl-4-(trifluoromethoxy)aniline
N-Ethyl-4-(trifluoromethoxy)aniline
Formula: C₉H₁₀F₃NO
MW: 205.18 g/mol
MDL Number: MFCD00203992
CAS Number: 175278-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR