N-Ethyl-2-(trifluoromethoxy)aniline
Supplier: Thermo Scientific
Synonyms:
2-(Trifluoromethoxy)ethylaminobenzene
TL00691.1
TL00691.10
MAYBTL00691.1EA
0
EUR
MAYBTL00691.1
MAYBTL00691.10
N-Ethyl-2-(trifluoromethoxy)aniline
N-Ethyl-2-(trifluoromethoxy)aniline
Formula:
C₉H₁₀F₃NO MW: 205.18 g/mol |
MDL Number:
MFCD00204184 CAS Number: 175278-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...