Azobenzene

Supplier: LGC Standards PROMOCHEM

PROOICRS0217EA 231 EUR
PROOICRS0217
Azobenzene
Azobenzene
Formula: C₁₂H₁₀N₂
MW: 182,22 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 66…69 °C
Density: 1,203 g/cm³ (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00003022
UN: 3077
ADR: 9,III
Merck Index: 12,00948

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR