Tricine
Supplier: Molekula
Synonyms:
2-((1,3-Dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid
, N-(Trishydroxymethylmethyl)glycine
MOLE16725607-500GEA
412
EUR
MOLE16725607-500G
Tricine
Tricine
Formula:
(HOCH₂)₃CNHCH₂CO₂H MW: 179,17 g/mol Melting Pt: 187 °C Density: 1,523 g/cm³ (19.85 °C) |
MDL Number:
MFCD00004277 CAS Number: 5704-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...