2-Amino-6-(trifluoromethyl)pyridine

Supplier: Molekula

MOLE52130800-1GEA 0 EUR
MOLE52130800-1G
2-Amino-6-(trifluoromethyl)pyridine
2-Amino-6-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162,11 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 78…80 °C
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR