(Benzyl)triphenylphosphonium chloride
Supplier: Molekula
MOLE26409937-100GEA
0
EUR
MOLE26409937-100G
(Benzyl)triphenylphosphonium chloride
(Benzyl)triphenylphosphonium chloride
Formula:
C₂₅H₂₂ClP MW: 388,88 g/mol Melting Pt: 345…347 °C Density: 1,18 g/cm³ (20 °C) |
MDL Number:
MFCD00011913 UN: 3464 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...