Oxalic acid ≥99%

Supplier: VWR Chemicals

20560.294EA 0 EUR
20560.294
Oxalic acid ≥99%
Oxalic acid
Formula: C₂H₂O₄
MW: 90,04 g/mol
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 99…100 °C
Density: 1,9 g/cm³ (20 °C)
MDL Number: MFCD00002573
UN: 3261
ADR: 8,III
Merck Index: 13,06980

Order Now

Specification Test Results

Assay Min. 99 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 0.05 %
Fe (Iron) Max. 20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR