Toluidine blue O

Supplier: Roth Carl

Synonyms: TBO, 3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chloride, 3-Amino-7-(dimethylamino)-2-methylphenothiazin-5-ium chloride, Tolonium chloride, Basic Blue 17, Blutene chloride

0300.1 0300.2
ROTH0300.1EA 25.3 EUR
ROTH0300.1 ROTH0300.2
Toluidine blue O
Toluidine blue O
Formula: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011934
CAS Number: 92-31-9
EINECS: 202-146-2
Merck Index: 12,09658

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR