N-Methylcyclohexylamine, Sigma-Aldrich®

Supplier: Merck

Synonyms: N-Cyclohexylmethylamine, Cyclohexyl(methyl)amine

M-Clarity™ quality level = MQ200

103322-100G 103322-5G
SIAL103322-100GEA 53.3 EUR
SIAL103322-100G SIAL103322-5G
N-Methylcyclohexylamine, Sigma-Aldrich®
N-Methylcyclohexylamine
Formula: C₇H₁₅N
MW: 113.2 g/mol
Boiling Pt: 149
Melting Pt: –8 °C
Density: 0.859 g/cm³ (20 °C)
MDL Number: MFCD00003832
CAS Number: 100-60-7
EINECS: 202-869-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR