Octadecyltrichlorosilane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Trichloro(stearyl)silane, Trichlorooctadecylsilane, Octadecyltrichlorosilane, ODTS, Trichloro(octadecyl)silane
M-Clarity™ quality level = MQ100
104817-100G 104817-25G 104817-500G
SIAL104817-100GEA 376 EUR
SIAL104817-100G SIAL104817-25G SIAL104817-500G
Octadecyltrichlorosilane, Sigma-Aldrich®
Octadecyltrichlorosilane
Formula: C₁₈H₃₇Cl₃Si
MW: 387.94 g/mol
Boiling Pt: 361 °C (1013 hPa)
Melting Pt: 22 °C
Density: 0.98 g/cm³ (20 °C)
Flash Pt: 89 °C
MDL Number: MFCD00000484
CAS Number: 112-04-9
EINECS: 203-930-7
UN: 1800
ADR: 8,II