Methyl cyanoacetate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Methyl 2-cyanoacetate
M-Clarity™ quality level = MQ500 (more info.)
108421-25G 108421-2KG 108421-500G
SIAL108421-25GEA 49.9 EUR
SIAL108421-25G SIAL108421-2KG SIAL108421-500G
Methyl cyanoacetate, Sigma-Aldrich®
Methyl cyanoacetate
Formula: C₄H₅NO₂
MW: 99.09 g/mol
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: –22.5 °C
Density: 1.13 g/cm³ (20 °C)
MDL Number: MFCD00001939
CAS Number: 105-34-0
EINECS: 203-288-8