Trimethyl orthoformate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
108456-20L 108456-250ML
SIAL108456-20LEA 1000 EUR
SIAL108456-20L SIAL108456-250ML
Trimethyl orthoformate, Sigma-Aldrich®
Trimethyl orthoformate
Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 101 °C (1013 hPa)
Melting Pt: –53 °C
Density: 0.9691 g/cm³ (20 °C)
Flash Pt: 13 °C
MDL Number: MFCD00008483
CAS Number: 149-73-5
EINECS: 205-745-7
UN: 3272
ADR: 3,II
Merck Index: 12,07012