Allylthiourea, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
N-Allylthiourea, Tiosinamine, 1-Allyl-2-thiourea, Allyl thiourea, Rhodalline, Thiosinamine, N-Allylthiourea
M-Clarity™ quality level = MQ100
108804-250G
108804-50G
SIAL108804-250GEA
191
EUR
SIAL108804-250G
SIAL108804-50G
Allylthiourea, Sigma-Aldrich®
Allylthiourea
Formula:
H₂CCHCH₂NHC(S)NH₂ MW: 116.19 g/mol Boiling Pt: 191 °C (1013 hPa) Melting Pt: 74 °C Density: 1.219 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004940 CAS Number: 109-57-9 EINECS: 203-683-5 UN: 2811 ADR: 6.1,III Merck Index: 12,09500 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...