2-Aminobenzothiazole, Sigma-Aldrich®
Supplier: Merck
Synonyms: Benzothiazol-2-ylamine, Benzo[d]thiazol-2-amine, Cerium(III)-Ionophore II
M-Clarity™ quality level = MQ100
108812-100G 108812-5G
SIAL108812-100GEA 68.3 EUR
SIAL108812-100G SIAL108812-5G
2-Aminobenzothiazole, Sigma-Aldrich®
2-Aminobenzothiazole
Formula: C₇H₆N₂S
MW: 150.2 g/mol
Boiling Pt: 190…195 °C (0.05 mmHg)
Melting Pt: 127…130 °C
MDL Number: MFCD00005785
CAS Number: 136-95-8
EINECS: 205-268-4
Merck Index: 13,00423