4-Chloro-o-phenylenediamine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1-Chloro-3,4-diaminobenzene, 4-Chlorobenzene-1,2-diamine, 4-Chloro-1,2-phenylenediamine, 4-Chloro-1,2-diaminobenzene
M-Clarity™ quality level = MQ100
108871-25G
108871-5G
SIAL108871-25GEA
96.7
EUR
SIAL108871-25G
SIAL108871-5G
4-Chloro-o-phenylenediamine, Sigma-Aldrich®
4-Chloro-o-phenylenediamine
Formula:
C₆H₇ClN₂ MW: 142.59 g/mol Boiling Pt: 280 °C (1013 hPa) Melting Pt: 89…90 °C |
MDL Number:
MFCD00011691 CAS Number: 95-83-0 EINECS: 202-456-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...