Methacrylamide, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
109606-250G
109606-500G
SIAL109606-250GEA
41.4
EUR
SIAL109606-250G
SIAL109606-500G
Methacrylamide, Sigma-Aldrich®
Methacrylamide
Formula:
C₄H₇NO MW: 85.11 g/mol Boiling Pt: 215 Melting Pt: 106…109 °C Density: 1.053 g/cm³ (20 °C) Flash Pt: 103 |
MDL Number:
MFCD00008018 CAS Number: 79-39-0 EINECS: 201-202-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...