1-Bromo-4-ethylbenzene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

111414-10G 111414-50G
SIAL111414-10GEA 63.1 EUR
SIAL111414-10G SIAL111414-50G
1-Bromo-4-ethylbenzene, Sigma-Aldrich®
1-Bromo-4-ethylbenzene
Formula: C₈H₉Br
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –43.5 °C
Density: 1.34226 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000111
CAS Number: 1585-07-5
EINECS: 216-439-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR