Ethyl salicylate, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Ethyl 2-hydroxybenzoate

112291-250G 112291-100G
SIAL112291-250GEA 48.7 EUR
SIAL112291-250G SIAL112291-100G
Ethyl salicylate, Sigma-Aldrich®
Ethyl salicylate
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: 1 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00002215
CAS Number: 118-61-6
EINECS: 204-265-5
Merck Index: 13,03881

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR