Crotonic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: trans-Crotonic acid, 2-Butenoic acid, trans-2-Butenoic acid
M-Clarity™ quality level = MQ100
113018-3KG 113018-500G
SIAL113018-3KGEA 268 EUR
SIAL113018-3KG SIAL113018-500G
Crotonic acid, Sigma-Aldrich®
Crotonic acid
Formula: C₄H₆O₂
Boiling Pt: 185…199 °C (1013 hPa)
Melting Pt: 71…72 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00002701
CAS Number: 107-93-7
EINECS: 203-533-9
UN: 2823
ADR: 8,III
Merck Index: 13,02625