4,4'-Difluorobenzophenone, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Bis(4-Fluorophenyl)methanone, Bis(4-fluorophenyl) ketone

M-Clarity™ quality level = MQ100

115495-100G 115495-25G
SIAL115495-100GEA 210 EUR
SIAL115495-100G SIAL115495-25G
4,4'-Difluorobenzophenone, Sigma-Aldrich®
4,4'-Difluorobenzophenone
Formula: C₁₃H₈F₂O
MW: 218.2 g/mol
Boiling Pt: 170…172 °C (10 mmHg)
Melting Pt: 140…141 °C
Density: 1.39 g/cm³ (20 °C)
MDL Number: MFCD00000353
CAS Number: 345-92-6
EINECS: 206-466-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR