9-Decen-1-ol, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Decen-10-ol

M-Clarity™ quality level = MQ100

118354-10G 118354-5G
SIAL118354-10GEA 69.2 EUR
SIAL118354-10G SIAL118354-5G
9-Decen-1-ol, Sigma-Aldrich®
9-Decen-1-ol
Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: –13 °C
Density: 0.8446 g/cm³ (20 °C)
MDL Number: MFCD00002992
CAS Number: 13019-22-2
EINECS: 235-878-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR