2-Deoxy-D(-)-ribose, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2-Deoxy-D(-)-erythropentose, Thyminose, 2-Deoxy-D(-)-arabinose, 2'-Deoxy-D-Ribose, (3S,4R)-3,4,5-Trihydroxypentanal
M-Clarity™ quality level = MQ100
121649-1G
121649-25G
121649-5G
SIAL121649-1GEA
28
EUR
SIAL121649-1G
SIAL121649-25G
SIAL121649-5G
2-Deoxy-D(-)-ribose, Sigma-Aldrich®
2-Deoxy-D(-)-ribose
Formula:
C₅H₁₀O₄ MW: 134.13 g/mol Boiling Pt: 292 °C (1013 hPa) Melting Pt: 95…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00135904 CAS Number: 533-67-5 EINECS: 208-573-0 Merck Index: 13,02925 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...