2-Aminothiazole, Sigma-Aldrich®

Supplier: Merck

Synonyms: Thiazol-2-ylamine, Aminothiazole, Thiazol-2-amine

M-Clarity™ quality level = MQ100

123129-100G 123129-500G 123129-5G
SIAL123129-100GEA 48.9 EUR
SIAL123129-100G SIAL123129-500G SIAL123129-5G
2-Aminothiazole, Sigma-Aldrich®
2-Aminothiazole
Formula: C₃H₄N₂S
MW: 100.14 g/mol
Boiling Pt: 216 °C (1013 hPa)
Melting Pt: 86…89 °C
Density: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00005325
CAS Number: 96-50-4
EINECS: 202-511-6
Merck Index: 13,00479

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR