Undec-10-enoic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Undecylenic-10 acid, Undecenoic acid, 10-Undecenoic acid, Undecylenic acid, Undecenylic acid
M-Clarity™ quality level = MQ200
124672-100ML
124672-500ML
SIAL124672-100MLEA
27.2
EUR
SIAL124672-100ML
SIAL124672-500ML
Undec-10-enoic acid, Sigma-Aldrich®
Undec-10-enoic acid
Formula:
C₁₁H₂₀O₂ MW: 184.28 g/mol Boiling Pt: 300 °C (1013 hPa) Melting Pt: 24 °C Density: 0.91 g/cm³ (20 °C) Flash Pt: 146 °C |
MDL Number:
MFCD00004442 CAS Number: 112-38-9 EINECS: 203-965-8 Merck Index: 12,09983 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...