N,N-Diethyl-1,1,1-trimethylsilylamine, Sigma-Aldrich®

Supplier: Merck

Synonyms: Trimethylsilyldiethylamine, (Diethylamino)trimethylsilane, N,N-Diethylaminotrimethylsilane, N,N-Diethyltrimethylsilylamine, N-(Trimethylsilyl)diethylamine, TMSDEA

M-Clarity™ quality level = MQ100

127256-100G 127256-25G
SIAL127256-100GEA 108 EUR
SIAL127256-100G SIAL127256-25G
N,N-Diethyl-1,1,1-trimethylsilylamine, Sigma-Aldrich®
N,N-Diethyl-1,1,1-trimethylsilylamine
Formula: C₇H₁₉NSi
MW: 145.32 g/mol
Boiling Pt: 126…127 °C (1013 hPa)
Melting Pt: –10 °C
Density: 0.7074 g/cm³ (20 °C)
Flash Pt: 15
Storage Temperature: Ambient
MDL Number: MFCD00009040
CAS Number: 996-50-9
EINECS: 213-637-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR