Ethyl benzoylacetate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Ethyl 3-oxo-3-phenylpropanoate

12990-100ML 12990-500ML
SIAL12990-100MLEA 65.6 EUR
SIAL12990-100ML SIAL12990-500ML
Ethyl benzoylacetate, Sigma-Aldrich®
Ethyl benzoylacetate
Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.114 g/cm³ (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00009196
CAS Number: 94-02-0
EINECS: 202-295-3
Merck Index: 13,03803

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR