2,2,2-Trichloroethyl chloroformate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
142077-25G 142077-100G
SIAL142077-25GEA 70.2 EUR
SIAL142077-25G SIAL142077-100G
2,2,2-Trichloroethyl chloroformate, Sigma-Aldrich®
2,2,2-Trichloroethyl chloroformate
Formula: C₃H₂Cl₄O₂
MW: 211.86 g/mol
Boiling Pt: 171
Melting Pt: 1
Density: 1.53 g/cm³ (20 °C)
MDL Number: MFCD00000810
CAS Number: 17341-93-4
EINECS: 241-363-7
UN: 3277
ADR: 6.1 II