2,3-Dimethyl-1,3-butadiene stabilised, Sigma-Aldrich®
Supplier: Merck
Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 100 ppm
M-Clarity™ quality level = MQ200
145491-10G 145491-50G
SIAL145491-10GEA 106 EUR
SIAL145491-10G SIAL145491-50G
2,3-Dimethyl-1,3-butadiene stabilised, Sigma-Aldrich®
2,3-Dimethyl-1,3-butadiene
Formula: C₆H₁₀
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –76 °C
Density: 0.7261 g/cm³ (20 °C)
MDL Number: MFCD00008595
CAS Number: 513-81-5
EINECS: 208-172-0
UN: 3295
ADR: 3,II
Merck Index: 13,03265