4-Pyridylmethanol, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 4-Pyridylmethanol, 4-(Hydroxymethyl)pyridine, 4-Pyridinemethanol, Pyridine-4-methanol, 4-Pyridylcarbinol

M-Clarity™ quality level = MQ100

151629-100G 151629-25G
SIAL151629-100GEA 131 EUR
SIAL151629-100G SIAL151629-25G
4-Pyridylmethanol, Sigma-Aldrich®
4-Pyridylmethanol
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 49…50 °C
Density: 1.15 g/cm³ (20 °C)
MDL Number: MFCD00006442
CAS Number: 586-95-8
EINECS: 209-590-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR