Isoxazole, Sigma-Aldrich®

Supplier: Merck


MQ200

151637-25G 151637-5G
SIAL151637-25GEA 80.2 EUR
SIAL151637-25G SIAL151637-5G
Isoxazole, Sigma-Aldrich®
Isoxazole
Formula: C₃H₃NO
MW: 69.06 g/mol
Boiling Pt: 95
Density: 1.08 g/cm³ (20 °C)
Flash Pt: 10
MDL Number: MFCD00003149
CAS Number: 288-14-2
EINECS: 206-018-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR