N,O-Bis(trimethylsilyl)trifluoroacetamide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate, Bis(Trimethylsilyl)Trifluoroacetamide, BSTFA
M-Clarity™ quality level = MQ200
155195-100G 155195-25G 155195-5G
SIAL155195-100GEA 340 EUR
SIAL155195-100G SIAL155195-25G SIAL155195-5G
N,O-Bis(trimethylsilyl)trifluoroacetamide, Sigma-Aldrich®
N,O-Bis(trimethylsilyl)trifluoroacetamide
Formula: C₈H₁₈F₃NOSi₂
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –41 °C
Density: 0.96 g/cm³ (20 °C)
Flash Pt: 34 °C
MDL Number: MFCD00008269
CAS Number: 25561-30-2
EINECS: 247-103-9
UN: 2924
ADR: 3,II